Anti-CCND1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77852-514
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CCND1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P24385
- Antigen synonyms:AI327039|B cell CLL/lymphoma 1|B cell leukemia 1|B cell lymphoma 1 protein|B-cell lymphoma 1 protein|BCL 1|BCL-1|BCL-1 oncogene|BCL1|BCL1 oncogene|ccnd1|CCND1/FSTL3 fusion gene|included|CCND1/IGHG1 fusion gene|CCND1/IGLC1 fusion gene|CCND1/PTH fusion gene|CCND1_HUMAN|cD1|Cyl 1|D11S287E|G1/S specific cyclin D1|G1/S-specific cyclin-D1|Parathyroid adenomatosis 1|PRAD1|PRAD1 oncogene|U21B31
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human G1/S-specific cyclin-D1 protein (1-295AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CCND1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human