Anti-FTSJ1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77855-637
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:FTSJ1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UET6
- Antigen synonyms:CDLIV|FTSJ 1|FtsJ homolog 1 (E. coli)|FtsJ homolog 1|FtsJ RNA methyltransferase homolog 1|FTSJ1|JM23|Mental retardation X linked 44|Mental retardation X linked 9|MRX44|MRX9|Protein ftsJ homolog 1|Putative ribosomal RNA methyltransferase 1|RRMJ1|RRMJ1_HUMAN|rRNA (uridine 2' O ) methyltransferase|rRNA (uridine-2''-O-)-methyltransferase|SPB1|TRM7
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Putative tRNA (cytidine(32)/guanosine(34)-2\\\'-O)-methyltransferase protein (1-329AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:20 to 1:200
Type:
Antigen: FTSJ1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse