Anti-APOBEC3A Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-294
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:APOBEC3A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P31941
- Antigen synonyms:A3A|ABC3A_HUMAN|Apolipoprotein B mRNA editing enzyme|catalytic polypeptide like 3A|ARP3|bK150C2.1|Catalytic polypeptide like 3A|Phorbolin 1|Phorbolin-1|PHRBN|Probable DNA dC >dU editing enzyme APOBEC-3A|Probable DNA dC->dU-editing enzyme APOBEC-3A
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human DNA dC->dU-editing enzyme APOBEC-3A protein (1-199AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: APOBEC3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human