Anti-TIMM10B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TIMM10B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-446
- 77845-445
- Antigen symbol:
- TIMM10B
- TIMM10B
- Antigen synonyms:
- Fracture callus 1 homolog (rat)|Fracture callus 1 homolog|Fracture callus protein 1|FxC1|Mitochondrial import inner membrane translocase subunit Tim10 B|Mitochondrial import inner membrane translocase subunit Tim9 B|OTTHUMP00000164584|OTTHUMP00000230720|TIM 10B|Tim10b|Tim9b|TIM9B_HUMAN|Translocase of inner mitochondrial membrane 10 homolog B (yeast)
- Fracture callus 1 homolog (rat)|Fracture callus 1 homolog|Fracture callus protein 1|FxC1|Mitochondrial import inner membrane translocase subunit Tim10 B|Mitochondrial import inner membrane translocase subunit Tim9 B|OTTHUMP00000164584|OTTHUMP00000230720|TIM 10B|Tim10b|Tim9b|TIM9B_HUMAN|Translocase of inner mitochondrial membrane 10 homolog B (yeast)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitochondrial import inner membrane translocase subunit Tim10 B protein (1-103AA)
- Recombinant Human Mitochondrial import inner membrane translocase subunit Tim10 B protein (1-103AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together