Human Synthetic CCR4 (from HEK293 Cells)
Full length Human CCR4, expressed in HEK293 cells, suitable for ELISA, SDS-PAGE.
: This product is for esearch use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilized: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P51679
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:C-C chemokine receptor type 4|C-C CKR-4|CC-CKR-4|CCR-4|CD194|CMKBR4|K5-5
- Protein/peptide name:CCR4
- Molecular weight:Full length human CCR4 protein has a MW of 41.4 kDa
- Concentration:Reconstitution dependent
- Formulation:Lyophilized from nanodisc solubilization buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8.0). Normally 5 - 8% trehalose is added as a protectant before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:ELISA, SDS-PAGE
Frequently Bought Together