Anti-ARL4C Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-389
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ARL4C
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P56559
- Antigen synonyms:ADP ribosylation factor like 4C|ADP ribosylation factor like 7|ADP-ribosylation factor-like protein 4C|ADP-ribosylation factor-like protein 7|ADP-ribosylation factor-like protein LAK|Arl4c|ARL4C_HUMAN|ARL7|LAK
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human ADP-ribosylation factor-like protein 4C protein (2-192AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IHC: 1:50 to 1:200
Type:
Antigen: ARL4C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human