Anti-ILVBL Rabbit Recombinant Polyclonal Antibody
Catalogue # 77858-097
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:ILVBL
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:A1L0T0
- Antigen synonyms:209L8|Acetolactate synthase homolog|Acetolactate synthase like protein|acetolactate synthase|bacterial|homolog of|Acetolactate synthase-like protein|AHAS|FLJ39061|ILV2H|IlvB (bacterial acetolactate synthase) like|IlvB like protein|IlvB-like protein|Ilvbl|ILVBL_HUMAN|MGC1269|MGC19535
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Acetolactate synthase-like protein (45-344AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: ILVBL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human