Anti-ENTPD5 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-764
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ENTPD5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O75356
- Antigen synonyms:CD39 like 4|AI196558|AI987697|CD39 antigen like 4|CD39 antigen-like 4|CD39L4|Ectonucleoside triphosphate diphosphohydrolase 5|ENTP5_HUMAN|Entpd5|ER UDPase|ER-UDPase|GDPase ENTPD5|Guanosine-diphosphatase ENTPD5|MGC163357|MGC163359|mNTPase|NTPDase 5|Nucleoside diphosphatase|PCPH|Pcph proto oncogene protein|Proto oncogene CPH|UDPase ENTPD5|Uridine-diphosphatase ENTPD5
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 5 protein (149-428AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: ENTPD5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse