Anti-CEP164 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CEP164
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-218
- 77845-219
- Antigen symbol:
- CEP164
- CEP164
- Antigen synonyms:
- AI450905|BC027092|CE164_HUMAN|Centrosomal protein 164|Centrosomal protein 164kDa|Centrosomal protein of 164 kDa|D030051D21|FLJ54767|KIAA1052|MGC38792|mKIAA1052|NPHP15|OTTMUSP00000042044|RGD1560988|RGD1561243
- AI450905|BC027092|CE164_HUMAN|Centrosomal protein 164|Centrosomal protein 164kDa|Centrosomal protein of 164 kDa|D030051D21|FLJ54767|KIAA1052|MGC38792|mKIAA1052|NPHP15|OTTMUSP00000042044|RGD1560988|RGD1561243
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UPV0
- Q9UPV0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Centrosomal protein of 164 kDa protein (1-112AA)
- Recombinant Human Centrosomal protein of 164 kDa protein (1-112AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together