Anti-SYNCRIP Rabbit Polyclonal Antibody
Catalog # 77848-425
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:SYNCRIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:O60506
- Antigen synonyms:cytoplasmic RNA-interacting protein|dJ3J17.2|Glycine and tyrosine rich RNA binding protein|Glycine- and tyrosine-rich RNA-binding protein|GRY RBP|GRY-RBP|GRYRBP|Heterogeneous nuclear ribonucleoprotein Q|hnRNP Q|HNRPQ|HNRPQ_HUMAN|HNRPQ1|NS1 associated protein 1|NS1-associated protein 1|NSAP1|pp68|RP1 3J17.2|Synaptotagmin binding cytoplasmic RNA interacting protein|Synaptotagmin-binding|Syncrip
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Internal of Human hnRNP Q.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000, IF: 1:100 to 1:500
Type:
Antigen: SYNCRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat