Order Entry
Canada
ContactUsLinkComponent
 
Anti-CHN2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77853-088
Supplier:  AFG Bioscience
New Product
undefined
Anti-CHN2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77853-088
Supplier:  AFG Bioscience
Supplier Number:  A17269-50UG
New Product

Specifications

  • Antigen symbol:
    CHN2
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    P52757
  • Antigen synonyms:
    ARHGAP3|BCH|Beta-chimaerin|Beta-chimerin|Rho GTPase-activating protein 3
  • Storage buffer:
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human Beta-chimaerin protein (1-91AA)
  • Purification:
    >95%, Protein G purified
  • Packaging:
    Vial

Specifications

About this item

Type:
Antigen: CHN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human