Anti-SUFU Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: SUFU
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77844-322
- 77844-321
- Antigen symbol:
- SUFU
- SUFU
- Antigen synonyms:
- OTTHUMP00000020374|OTTHUMP00000020377|OTTHUMP00000020379|PRO1280|SU FU|SU(F)U|Su(fu)|SUFU negative regulator of hedgehog signaling|SUFU_HUMAN|SUFUH|SUFUXL|Suppressor of Fused|Suppressor of fused homolog (Drosophila)|Suppressor of fused homolog
- OTTHUMP00000020374|OTTHUMP00000020377|OTTHUMP00000020379|PRO1280|SU FU|SU(F)U|Su(fu)|SUFU negative regulator of hedgehog signaling|SUFU_HUMAN|SUFUH|SUFUXL|Suppressor of Fused|Suppressor of fused homolog (Drosophila)|Suppressor of fused homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UMX1
- Q9UMX1
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Suppressor of fused homolog protein (325-484AA)
- Recombinant Human Suppressor of fused homolog protein (325-484AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together