Anti-COX6B1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77852-123
New Product
- Antigen symbol:COX6B1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P14854
- Antigen synonyms:COX 6B|COX VIb 1|COX VIb-1|COX6B|COXG|COXVIb1|CX6B1_HUMAN|Cytochrome c oxidase subunit 6B1|Cytochrome c oxidase subunit VIb|Cytochrome c oxidase subunit VIb isoform 1|cytochrome c oxidase subunit VIb polypeptide 1 (ubiquitous)|Cytochrome c oxidase subunit Vib polypeptide 1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cytochrome c oxidase subunit 6B1 protein (1-86AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: COX6B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human