Anti-AGGF1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-868
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:AGGF1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:Q8N302
- Antigen synonyms:AGGF 1|AGGF1_HUMAN|Angiogenic factor VG5Q|Angiogenic factor with G patch and FHA domains 1|G patch domain containing protein 7|G patch domain-containing protein 7|GPATC 7|GPATC7|GPATCH 7|GPATCH7|HSU84971|HUS84971|hVG5Q|Vasculogenesis gene on 5q|Vasculogenesis gene on 5q protein|VG5Q
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Angiogenic factor with G patch and FHA domains 1 protein (1-70AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human