Anti-CCHCR1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-479
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CCHCR1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:Q8TD31
- Antigen synonyms:C6orf18|HCR|Coiled-coil alpha-helical rod protein 1|Alpha-helical coiled-coil rod protein|Putative gene 8 protein|Pg8
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Coiled-coil alpha-helical rod protein 1 protein (483-782AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CCHCR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human