Recombinant Human IL-8 (from HEK293 cells)
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Frequently Bought Together
Alconox® Powdered Precision Cleaners
Supplier: Alconox
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
Supplier: Micro Essential
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
HisTag® Antibody HRP Conjugate Kit, MilliporeSigma
Supplier: MilliporeSigma
Difco™ M17 Broth, BD Diagnostics
Supplier: BD
Manganese(II) Chloride
Supplier: Ward's Science
Water USP, Ph. Eur., purified
Supplier: Ricca Chemical
QUICK-LOAD® Taq 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
GASTIGHT® 1000 Series Syringes, Hamilton
Supplier: Hamilton
INDICATOR PAPER WATOR DARK BLUE 78X40 MM
Supplier: Macherey-Nagel