Anti-EEF1A2 Rabbit Recombinant Polyclonal Antibody
77854-506
New Product
- Antigen symbol:EEF1A2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:Q05639
- Antigen synonyms:eEF1A-2|EEF1AL|EF 1 alpha 2|EF-1-alpha-2|EF1A|EF1A2_HUMAN|Elongation factor 1 A 2|Elongation factor 1 alpha|Elongation factor 1 alpha 2|Elongation factor 1-alpha 2|Eukaryotic elongation factor 1 A-2|Eukaryotic translation elongation factor 1 alpha 2|FLJ41696|HS1|OTTHUMP00000031776|Statin like|Statin S1|Statin-S1|STN|STNL
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human Elongation factor 1-alpha 2 protein (184-463AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: EEF1A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human