Anti-IL1RL2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IL1RL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-256
- 77843-255
- Antigen symbol:
- IL1RL2
- IL1RL2
- Antigen synonyms:
- IL 1 receptor related protein 2|IL 1Rrp2|IL 36R|IL-1Rrp2|IL-36 receptor|IL1 receptor related protein 2|IL1R rp2|IL1R-rp2|IL1RRP2|IL36R|ILRL2_HUMAN|Interleukin 1 receptor like 2|Interleukin 1 receptor related protein 2|Interleukin-1 receptor-like 2|Interleukin-1 receptor-related protein 2
- IL 1 receptor related protein 2|IL 1Rrp2|IL 36R|IL-1Rrp2|IL-36 receptor|IL1 receptor related protein 2|IL1R rp2|IL1R-rp2|IL1RRP2|IL36R|ILRL2_HUMAN|Interleukin 1 receptor like 2|Interleukin 1 receptor related protein 2|Interleukin-1 receptor-like 2|Interleukin-1 receptor-related protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HB29
- Q9HB29
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interleukin-1 receptor-like 2 protein (20-218AA)
- Recombinant Human Interleukin-1 receptor-like 2 protein (20-218AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together