Anti-NUP98 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NUP98
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-242
- 77745-241
- Antigen symbol:
- NUP98
- NUP98
- Antigen synonyms:
- 96 kDa nucleoporin|98 kDa nucleoporin|ADAR2|ADIR2|GLFG-repeat containing nucleoporin|Nuclear pore complex protein Nup96|Nuclear pore complex protein Nup98 Nup96|Nucleoporin 98kD|nucleoporin 98kDa|Nucleoporin Nup96|Nucleoporin Nup98|NUP196|NUP96|Nup98|Nup98-Nup96|NUP98_HUMAN
- 96 kDa nucleoporin|98 kDa nucleoporin|ADAR2|ADIR2|GLFG-repeat containing nucleoporin|Nuclear pore complex protein Nup96|Nuclear pore complex protein Nup98 Nup96|Nucleoporin 98kD|nucleoporin 98kDa|Nucleoporin Nup96|Nucleoporin Nup98|NUP196|NUP96|Nup98|Nup98-Nup96|NUP98_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52948
- P52948
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nuclear pore complex protein Nup98-Nup96 protein (965-1110AA)
- Nuclear pore complex protein Nup98-Nup96 protein (965-1110AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together