Anti-MRPL14 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: MRPL14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77859-949
- 77743-393
- Antigen symbol:
- MRPL14
- MRPL14
- Antigen synonyms:
- 1110006I11Rik|39S ribosomal protein L14|39S ribosomal protein L14 mitochondrial|39S ribosomal protein L32|AI846816|L14mt|L32mt|mitochondrial|MRP L14|MRP L32|MRP-L14|MRP-L32|mrpl14|RM14_HUMAN
- 1110006I11Rik|39S ribosomal protein L14|39S ribosomal protein L14 mitochondrial|39S ribosomal protein L32|AI846816|L14mt|L32mt|mitochondrial|MRP L14|MRP L32|MRP-L14|MRP-L32|mrpl14|RM14_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P1L8
- Q6P1L8
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human 39S ribosomal protein L14, mitochondrial protein (1-145AA)
- Recombinant Human 39S ribosomal protein L14, mitochondrial protein (1-145AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together