Anti-MGAT2 Rabbit Polyclonal Antibody
Catalog # 77743-086
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MGAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q10469
- Antigen synonyms:MGAT2Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase|EC 2.4.1.143|Beta-1,2-N-acetylglucosaminyltransferase II|GlcNAc-T II|GNT-II|Mannoside acetylglucosaminyltransferase 2|N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MGAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat