Anti-JUN Rabbit Polyclonal Antibody
Catalog # 77740-666
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:JUN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P05412
- Antigen synonyms:Activator protein 1|AP 1|AP-1|AP1|cJun|Enhancer Binding Protein AP1|Jun Activation Domain Binding Protein|JUN|Jun oncogene|JUN protein|Jun proto oncogene|JUN_HUMAN|JUNC|Oncogene JUN|p39|Proto oncogene c jun|Proto oncogene cJun|Proto-oncogene c-jun|Transcription Factor AP 1|Transcription factor AP-1|Transcription Factor AP1|V jun avian sarcoma virus 17 oncogene homolog|V jun sarcoma virus 17 oncogene homolog (avian)|V jun sarcoma virus 17 oncogene homolog|V-jun avian sarcoma virus 17 oncogene homolog|vJun Avian Sarcoma Virus 17 Oncogene Homolog
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Activator protein 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: JUN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat