18403
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-751
- 77743-124
- Antigen synonyms:
- HOXB1|HOX2I|Homeobox protein Hox-B1|Homeobox protein Hox-2I|HOXA1|HOX1F|Homeobox protein Hox-A1|Homeobox protein Hox-1F|HOXD1|HOX4|HOX4G|Homeobox protein Hox-D1|Homeobox protein Hox-GG
- Gene ID:
- P14653/P49639/Q9GZZ0
- Immunogen:
- HOXB1
- Form:
- Liquid
- Purification:
- Antigen affinity purified
- Storage buffer:
- PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
Specifications
Frequently Bought Together