Anti-LTA4H Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: LTA4H
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77742-444
- 77859-210
- Antigen symbol:
- LTA4H
- LTA4H
- Antigen synonyms:
- FLJ17564|Leukotriene A(4) hydrolase|Leukotriene A-4 hydrolase|Leukotriene A4 hydrolase|LKHA4_HUMAN|LTA-4 hydrolase|LTA4|LTA4 hydrolase|LTA4H
- LTA4 hydrolase|LKHA4_HUMAN|FLJ17564|Leukotriene A-4 hydrolase|Leukotriene A(4) hydrolase|LTA4H|LTA4|Leukotriene A4 hydrolase|LTA-4 hydrolase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09960
- P09960
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leukotriene A-4 hydrolase protein (107-311AA)
- Recombinant Human Leukotriene A-4 hydrolase protein (107-311AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together