Anti-KPNA5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: KPNA5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-704
- 77741-826
- Antigen symbol:
- KPNA5
- KPNA5
- Antigen synonyms:
- IMA5_HUMAN|Importin alpha 6|Importin subunit alpha 6|Importin subunit alpha-6|IPOA 6|IPOA6|Karyopherin alpha 5|Karyopherin alpha 5 importin alpha 6|Karyopherin subunit alpha 5|Karyopherin subunit alpha-5|KPNA 5|SRP 6|SRP6
- IMA5_HUMAN|Importin alpha 6|Importin subunit alpha 6|Importin subunit alpha-6|IPOA 6|IPOA6|Karyopherin alpha 5|Karyopherin alpha 5 importin alpha 6|Karyopherin subunit alpha 5|Karyopherin subunit alpha-5|KPNA 5|SRP 6|SRP6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15131
- O15131
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Importin subunit alpha-6 protein (1-240AA)
- Recombinant Human Importin subunit alpha-6 protein (1-240AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together