Anti-HOXB2 Rabbit Polyclonal Antibody
Catalog # 77737-968
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HOXB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P14652
- Antigen synonyms:homeo box 2H| homeo box B2| homeobox B2| Homeobox protein Hox-2.8| Homeobox protein Hox-2H| Homeobox protein Hox-B2| Hox 2 8| Hox 2H| HOX2| HOX2H| HXB2_HUMAN| K8| K8 home protein
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:homeo box 2H
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:20000
Type:
Antigen: HOXB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human