Anti-MLLT10 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MLLT10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-816
- 77743-208
- Antigen symbol:
- MLLT10
- MLLT10
- Antigen synonyms:
- AF 10|AF10|AF10_HUMAN|ALL 1 fused gene from chromosome 10|ALL1 fused gene from chromosome 10|ALL1-fused gene from chromosome 10 protein|DKFZp686E10210|MGC75086|MLLT 10|Myeloid/lymphoid or mixed lineage leukemia (trithorax homolog Drosophila) translocated to 10|Myeloid/lymphoid or mixed lineage leukemia translocated to 10|OTTHUMP00000045273|Protein AF 10|Protein AF-10|Protein AF10|Type I AF10 protein
- AF 10|AF10|AF10_HUMAN|ALL 1 fused gene from chromosome 10|ALL1 fused gene from chromosome 10|ALL1-fused gene from chromosome 10 protein|DKFZp686E10210|MGC75086|MLLT 10|Myeloid/lymphoid or mixed lineage leukemia (trithorax homolog Drosophila) translocated to 10|Myeloid/lymphoid or mixed lineage leukemia translocated to 10|OTTHUMP00000045273|Protein AF 10|Protein AF-10|Protein AF10|Type I AF10 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55197
- P55197
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Protein AF-10 protein (1-126AA)
- Recombinant human Protein AF-10 protein (1-126AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together