Anti-NFYB Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NFYB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-405
- 77744-404
- Antigen symbol:
- NFYB
- NFYB
- Antigen synonyms:
- CAAT box DNA binding protein subunit B|CAAT box DNA-binding protein subunit B|CBF A|CBF B|CCAAT binding transcription factor subunit A|HAP 3|HAP3|NF Y protein chain B|NF YB|NF-YB|NFYB_HUMAN|Nuclear transcription factor Y beta|Nuclear transcription factor Y subunit B|Nuclear transcription factor Y subunit beta|Transcription factor NF Y B subunit
- CAAT box DNA binding protein subunit B|CAAT box DNA-binding protein subunit B|CBF A|CBF B|CCAAT binding transcription factor subunit A|HAP 3|HAP3|NF Y protein chain B|NF YB|NF-YB|NFYB_HUMAN|Nuclear transcription factor Y beta|Nuclear transcription factor Y subunit B|Nuclear transcription factor Y subunit beta|Transcription factor NF Y B subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25208
- P25208
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Nuclear transcription factor Y subunit beta protein (1-207AA)
- Nuclear transcription factor Y subunit beta protein (1-207AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together