Anti-IST1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: IST1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-571
- 77858-501
- Antigen symbol:
- IST1
- IST1
- Antigen synonyms:
- hIncreased sodium tolerance 1|Increased sodium tolerance 1 homolog|IST1 homolog|IST1_HUMAN|KIAA0174|MAPK activating protein PM28|MGC117220|MGC128433|Putative MAPK-activating protein PM28
- hIncreased sodium tolerance 1|Increased sodium tolerance 1 homolog|IST1 homolog|IST1_HUMAN|KIAA0174|MAPK activating protein PM28|MGC117220|MGC128433|Putative MAPK-activating protein PM28
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53990
- P53990
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human IST1 homolog protein (172-284AA)
- Recombinant human IST1 homolog protein (172-284AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together