Anti-EMX2 Rabbit Polyclonal Antibody
77736-523
New Product
- Antigen symbol:EMX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q04743
- Antigen synonyms:Empty spiracles homeobox 2|Empty spiracles homolog 2 (Drosophila)|Empty spiracles homolog 2|Empty spiracles like protein 2|Empty spiracles-like protein 2|EMX 2|EMX2_HUMAN|Homeobox protein EMX 2|Homeobox protein EMX2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Empty spiracles homeobox 2
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: EMX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse