Anti-HNF4A Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HNF4A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-510
- 77740-215
- Antigen symbol:
- HNF4A
- Antigen synonyms:
- FLJ39654|FRTS4|Hepatic nuclear factor 4 alpha|Hepatocyte nuclear factor 4 alpha|Hepatocyte nuclear factor 4|Hepatocyte nuclear factor 4-alpha|HNF 4 alpha|HNF 4|HNF-4-alpha|HNF4|HNF4A|HNF4A_HUMAN|HNF4a7|HNF4a8|HNF4a9|Hnf4alpha|HNF4alpha10/11/12|MODY 1|MODY|MODY1|NR2A1|NR2A21|Nuclear receptor subfamily 2 group A member 1|OTTHUMP00000031060|OTTHUMP00000031062|TCF 14|TCF|TCF-14|TCF14|Tcf4|Transcription factor 14|hepatic nuclear factor|Transcription factor HNF 4|Transcription factor HNF-4|Transcription factor HNF4
- FLJ39654|FRTS4|Hepatic nuclear factor 4 alpha|Hepatocyte nuclear factor 4 alpha|Hepatocyte nuclear factor 4|Hepatocyte nuclear factor 4-alpha|HNF 4 alpha|HNF 4|HNF-4-alpha|HNF4|HNF4A|HNF4A_HUMAN|HNF4a7|HNF4a8|HNF4a9|Hnf4alpha|HNF4alpha10/11/12|MODY 1|MODY|MODY1|NR2A1|NR2A21|Nuclear receptor subfamily 2 group A member 1|OTTHUMP00000031060|OTTHUMP00000031062|TCF 14|TCF|TCF-14|TCF14|Tcf4|Transcription factor 14|hepatic nuclear factor|Transcription factor HNF 4|Transcription factor HNF-4|Transcription factor HNF4
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P41235
- P41235
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Hepatocyte nuclear factor 4-alpha protein (130-330AA)
- Recombinant Human Hepatocyte nuclear factor 4-alpha protein (130-330AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together