Anti-LSS Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: LSS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-205
- 77742-437
- Antigen symbol:
- LSS
- Antigen synonyms:
- LSS|OSCLanosterol synthase|EC 5.4.99.7|2|3-epoxysqualene--lanosterol cyclase|Oxidosqualene--lanosterol cyclase|OSC|hOSC
- LSS|OSCLanosterol synthase|EC 5.4.99.7|2|3-epoxysqualene--lanosterol cyclase|Oxidosqualene--lanosterol cyclase|OSC|hOSC
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P48449
- P48449
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Lanosterol synthase protein (1-300AA)
- Recombinant Human Lanosterol synthase protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together