Anti-LONP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LONP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-055
- 77742-268
- Antigen symbol:
- LONP1
- Antigen synonyms:
- hLON|hLON ATP dependent protease|LON|lon peptidase 1|mitochondrial|LON protease|Lon protease homolog|Lon protease like protein|Lon protease-like protein|LONHs|LONM_HUMAN|LONP|Lonp1|MGC1498|Mitochondrial ATP dependent protease Lon|Mitochondrial ATP-dependent protease Lon|Mitochondrial lon peptidase 1|PIM1|Protease serine 15|PRSS15|Serine protease 15
- hLON|hLON ATP dependent protease|LON|lon peptidase 1|mitochondrial|LON protease|Lon protease homolog|Lon protease like protein|Lon protease-like protein|LONHs|LONM_HUMAN|LONP|Lonp1|MGC1498|Mitochondrial ATP dependent protease Lon|Mitochondrial ATP-dependent protease Lon|Mitochondrial lon peptidase 1|PIM1|Protease serine 15|PRSS15|Serine protease 15
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P36776
- P36776
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Lon protease homolog, mitochondrial protein (614-959AA)
- Recombinant Human Lon protease homolog, mitochondrial protein (614-959AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together