Anti-GREM1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GREM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-997
- 77856-433
- Antigen symbol:
- GREM1
- GREM1
- Antigen synonyms:
- BMP antagonist 1|Cell proliferation inducing gene 2 protein|Cell proliferation-inducing gene 2 protein|CKTSF1B1|colorectal adenoma and carcinoma 1|Cysteine knot superfamily 1|Cysteine knot superfamily BMP antagonist 1|DAN domain family member 2|DAND2|Down regulated in Mos-transformed cells protein|Down-regulated in Mos-transformed cells protein|DRM|GREM1_HUMAN|Gremlin 1 homolog|cysteine knot superfamily|Gremlin 1 like protein|Gremlin 1|homolog|GREMLIN|Gremlin-1|Gremlin1|IHG-2|IHG2|Increased in high glucose 2|Increased in high glucose protein 2|PIG2|Proliferation inducing gene 2|proliferation inducing gene 2 protein
- BMP antagonist 1|Cell proliferation inducing gene 2 protein|Cell proliferation-inducing gene 2 protein|CKTSF1B1|colorectal adenoma and carcinoma 1|Cysteine knot superfamily 1|Cysteine knot superfamily BMP antagonist 1|DAN domain family member 2|DAND2|Down regulated in Mos-transformed cells protein|Down-regulated in Mos-transformed cells protein|DRM|GREM1_HUMAN|Gremlin 1 homolog|cysteine knot superfamily|Gremlin 1 like protein|Gremlin 1|homolog|GREMLIN|Gremlin-1|Gremlin1|IHG-2|IHG2|Increased in high glucose 2|Increased in high glucose protein 2|PIG2|Proliferation inducing gene 2|proliferation inducing gene 2 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60565
- O60565
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Gremlin-1 protein (18-184AA)
- Recombinant Human Gremlin-1 protein (18-184AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together