Anti-MRPS5 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: MRPS5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77860-000
- 77743-482
- Antigen symbol:
- MRPS5
- Antigen synonyms:
- 28S ribosomal protein S5|Mitochondrial 28S ribosomal protein S5|mitochondrial|mitochondrial ribosomal protein S5|MRP S5|MRP-S5|Mrps5|RT05_HUMAN|S5mt
- 28S ribosomal protein S5|Mitochondrial 28S ribosomal protein S5|mitochondrial|mitochondrial ribosomal protein S5|MRP S5|MRP-S5|Mrps5|RT05_HUMAN|S5mt
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P82675
- P82675
- Isotype:
- IgG
- Western blot:
- Yes
- ELISA:
- Yes
- Immunogen:
- Recombinant Human 28S ribosomal protein S5, mitochondrial protein (161-430AA)
- Recombinant Human 28S ribosomal protein S5, mitochondrial protein (161-430AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together