Anti-MAT2A Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: MAT2A
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-815
- 77859-512
- Antigen symbol:
- MAT2A
- MAT2A
- Antigen synonyms:
- AdoMet synthase 2| AdoMet synthetase 2| AdoMet synthetase| AMS 2| AMS2| MAT 2| MAT 2A| MAT II| MAT-II| MATA 2| MATA2| MATII | Methionine adenosyltransferase 2| Methionine adenosyltransferase| Methionine adenosyltransferase II alpha| Methionine adenosyltransferase II| METK2_HUMAN| S adenosylmethionine synthetase gamma form| S adenosylmethionine synthetase isoform type 2| S-adenosylmethionine synthase isoform type-2| SAMS 2| SAMS2
- AdoMet synthase 2|AdoMet synthetase 2|AdoMet synthetase|AMS 2|AMS2|MAT 2|MAT 2A|MAT II|MAT-II|MATA 2|MATA2|MATII |Methionine adenosyltransferase 2|Methionine adenosyltransferase|Methionine adenosyltransferase II alpha|Methionine adenosyltransferase II|METK2_HUMAN|S adenosylmethionine synthetase gamma form|S adenosylmethionine synthetase isoform type 2|S-adenosylmethionine synthase isoform type-2|SAMS 2|SAMS2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31153
- P31153
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human S-adenosylmethionine synthase isoform type-2 protein (1-395AA)
- Recombinant Human S-adenosylmethionine synthase isoform type-2 protein (1-395AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together