Anti-LIN7C Rabbit Polyclonal Antibody
Catalog # 77742-217
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:LIN7C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:Q9NUP9
- Antigen synonyms:hLin 7|hLin7|LIN 7|Lin 7 homolog A|LIN 7A|LIN 7B|LIN 7C|Lin-7C|Lin7 homolog A|LIN7A|LIN7C|LIN7C_HUMAN|MALS 1|MALS|MALS-3|MALS1|Mammalian LIN 7 1|Mammalian lin seven protein 1|Mammalian lin-seven protein 3|Mammalian LIN7 1|MGC148143|Protein lin 7 homolog B|Protein lin 7 homolog C|Protein lin-7 homolog C|protein lin7 homolog A|Tax interaction protein 33|TIP 33|TIP33|VELI 1|Veli 1 protein|Veli-3|VELI1|Velis|Vertebrate LIN 7 homolog 1|Vertebrate lin-7 homolog 3|Vertebrate LIN7 homolog 1
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Vertebrate LIN7 homolog 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: LIN7C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat