Anti-MED12L Rabbit Polyclonal Antibody
77742-950
New Product
- Antigen symbol:MED12L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:Q86YW9
- Antigen synonyms:MED12L|KIAA1635|TNRC11L|TRALP|TRALPUSH|PRO0314|Mediator of RNA polymerase II transcription subunit 12-like protein|Mediator complex subunit 12-like protein|Thyroid hormone receptor-associated-like protein|Trinucleotide repeat-containing gene 11 protein-like
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Trinucleotide repeat-containing gene 11 protein-like
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: MED12L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat