Anti-LRRTM2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LRRTM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-393
- 77859-170
- Antigen symbol:
- LRRTM2
- LRRTM2
- Antigen synonyms:
- Leucine rich repeat transmembrane neuronal 2|FLJ99831|KIAA0416|Leucine rich repeat transmembrane neuronal 2 protein|LRRT2_HUMAN|LRRN2|FLJ76283|Leucine-rich repeat neuronal 2 protein|Leucine rich repeat transmembrane protein 2|Leucine-rich repeat transmembrane neuronal protein 2
- FLJ76283|FLJ99831|KIAA0416|Leucine rich repeat transmembrane neuronal 2|Leucine rich repeat transmembrane neuronal 2 protein|Leucine rich repeat transmembrane protein 2|Leucine-rich repeat neuronal 2 protein|Leucine-rich repeat transmembrane neuronal protein 2|LRRN2|LRRT2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43300
- O43300
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leucine-rich repeat transmembrane neuronal protein 2 protein (165-422AA)
- Recombinant Human Leucine-rich repeat transmembrane neuronal protein 2 protein (165-422AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together