Anti-GZMB Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GZMB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-324
- 77856-648
- Antigen symbol:
- GZMB
- GZMB
- Antigen synonyms:
- C11|Cathepsin G like 1|Cathepsin G-like 1|CCPI|CGL 1|CGL1|CSP B|CSPB|CTLA 1|CTLA-1|CTLA1|CTSGL1|Cytotoxic serine protease B|Cytotoxic T lymphocyte associated serine esterase 1|Cytotoxic T lymphocyte proteinase 2|Cytotoxic T-lymphocyte proteinase 2|Fragmentin 2|Fragmentin-2|GRAB_HUMAN|Granzyme 2|Granzyme B (granzyme 2|cytotoxic T lymphocyte associated serine esterase 1)|Granzyme B|Granzyme-2|GranzymeB|GRB|Hlp|Human lymphocyte protein|Lymphocyte protease|Protease|serine|B|SECT|T cell serine protease 1 3E|T cell serine protease 1-3E|T-cell serine protease 1-3E
- C11|Cathepsin G like 1|Cathepsin G-like 1|CCPI|CGL 1|CGL1|CSP B|CSPB|CTLA 1|CTLA-1|CTLA1|CTSGL1|Cytotoxic serine protease B|Cytotoxic T lymphocyte associated serine esterase 1|Cytotoxic T lymphocyte proteinase 2|Cytotoxic T-lymphocyte proteinase 2|Fragmentin 2|Fragmentin-2|GRAB_HUMAN|Granzyme 2|Granzyme B (granzyme 2|cytotoxic T lymphocyte associated serine esterase 1)|Granzyme B|Granzyme-2|GranzymeB|GRB|Hlp|Human lymphocyte protein|Lymphocyte protease|Protease|serine|B|SECT|T cell serine protease 1 3E|T cell serine protease 1-3E|T-cell serine protease 1-3E
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10144
- P10144
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Granzyme B protein (21-247AA)
- Recombinant Human Granzyme B protein (21-247AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together