Anti-Gpi Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: Gpi
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- 77738-754
- 77856-315
- Antigen symbol:
- Gpi
- Gpi
- Antigen synonyms:
- Gpi|Gpi1Glucose-6-phosphate isomerase|GPI|EC 5.3.1.9|Autocrine motility factor|AMF|Neuroleukin|NLK|Phosphoglucose isomerase|PGI|Phosphohexose isomerase|PHI
- Gpi|Neuroleukin|NLK|Gpi1Glucose-6-phosphate isomerase|Autocrine motility factor|AMF|EC 5.3.1.9|Phosphoglucose isomerase|PGI|Phosphohexose isomerase|PHI|GPI
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P06745
- P06745
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Mouse Glucose-6-phosphate isomerase protein (2-558AA)
- Recombinant Mouse Glucose-6-phosphate isomerase protein (2-558AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together