Anti-GIT1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GIT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-282
- 77856-008
- Antigen symbol:
- GIT1
- GIT1
- Antigen synonyms:
- ARF GAP ARF GTPase activating protein ARF GTPase-activating protein CAT 1|CAT-1|CaT1|Cool associated and tyrosine phosphorylated protein 1|Cool-associated and tyrosine-phosphorylated protein 1|G protein coupled receptor kinase interacting ArfGAP 1|G protein coupled receptor kinase interactor 1|G protein-coupled receptor kinase-interactor 1|GIT1_HUMAN|GRK interacting protein 1|GRK-interacting protein 1
- ARF GAP ARF GTPase activating protein ARF GTPase-activating protein CAT 1|CAT-1|CaT1|Cool associated and tyrosine phosphorylated protein 1|Cool-associated and tyrosine-phosphorylated protein 1|G protein coupled receptor kinase interacting ArfGAP 1|G protein coupled receptor kinase interactor 1|G protein-coupled receptor kinase-interactor 1|GIT1_HUMAN|GRK interacting protein 1|GRK-interacting protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2X7
- Q9Y2X7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human ARF GTPase-activating protein GIT1 protein (470-642AA)
- Recombinant Human ARF GTPase-activating protein GIT1 protein (470-642AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together