Anti-MTHFS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: MTHFS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77743-597
- 77860-095
- Antigen symbol:
- MTHFS
- MTHFS
- Antigen synonyms:
- 10-methenyl-tetrahydrofolate synthetase|5 10 methenyl tetrahydrofolate synthetase|5|5 formyltetrahydrofolate cyclo ligase|5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)|5-formyltetrahydrofolate cyclo-ligase|FLJ30410|HsT19268|Methenyl THF synthetase|Methenyl-THF synthetase|MTHFS|MTHFS_HUMAN
- 10-methenyl-tetrahydrofolate synthetase| 5 10 methenyl tetrahydrofolate synthetase| 5| 5 formyltetrahydrofolate cyclo ligase| 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)| 5-formyltetrahydrofolate cyclo-ligase| FLJ30410| HsT19268| Methenyl THF synthetase| Methenyl-THF synthetase| MTHFS| MTHFS_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49914
- P49914
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 5-formyltetrahydrofolate cyclo-ligase protein (2-203AA)
- Recombinant Human 5-formyltetrahydrofolate cyclo-ligase protein (2-203AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together