Anti-NDC80 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NDC80
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-103
- 77744-102
- Antigen symbol:
- NDC80
- NDC80
- Antigen synonyms:
- from Entrez Gene|HEC|Highly expressed in cancer|Highly expressed in cancer protein|Highly expressed in cancer rich in leucine heptad repeats|HsHec1|hsKinetochore associated 2|Kinetochore associated protein 2|Kinetochore protein Hec1|Kinetochore protein NDC80 homolog|Kinetochore-associated protein 2|KNTC2|NDC80 homolog kinetochore complex component|NDC80 kinetochore complex component homolog|S. cerevisiae|homolog of|NDC80_HUMAN|Retinoblastoma associated protein HEC|Retinoblastoma-associated protein HEC|TID3
- from Entrez Gene|HEC|Highly expressed in cancer|Highly expressed in cancer protein|Highly expressed in cancer rich in leucine heptad repeats|HsHec1|hsKinetochore associated 2|Kinetochore associated protein 2|Kinetochore protein Hec1|Kinetochore protein NDC80 homolog|Kinetochore-associated protein 2|KNTC2|NDC80 homolog kinetochore complex component|NDC80 kinetochore complex component homolog|S. cerevisiae|homolog of|NDC80_HUMAN|Retinoblastoma associated protein HEC|Retinoblastoma-associated protein HEC|TID3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14777
- O14777
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Kinetochore protein NDC80 homolog protein (1-249AA)
- Kinetochore protein NDC80 homolog protein (1-249AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together