Order Entry
Canada
ContactUsLinkComponent
Anti-NCS1 Rabbit Recombinant Polyclonal Antibody
Anti-NCS1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-NCS1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77855-609
  • 77737-621
  • Antigen symbol:
  • NCS1
  • NCS1
  • Antigen synonyms:
  • 9430075O15Rik|A730032G13Rik|AI836659|DKFZp761L1223|FLUP|FREQ|Frequenin|Frequenin homolog (Drosophila)|Frequenin homolog|Frequenin like protein|Drosophila|homolog of|Frequenin-like protein|Frequenin-like ubiquitous protein|Mfreq|NCS 1|NCS-1|NCS1_HUMAN|Neuronal calcium sensor 1|Neuronal Calicum Sensor 1
  • 9430075O15Rik|A730032G13Rik|AI836659|DKFZp761L1223|FLUP|FREQ|Frequenin|Frequenin homolog (Drosophila)|Frequenin homolog|Frequenin like protein|Drosophila|homolog of|Frequenin-like protein|Frequenin-like ubiquitous protein|Mfreq|NCS 1|NCS-1|NCS1_HUMAN|Neuronal calcium sensor 1|Neuronal Calicum Sensor 1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse, Rat
  • Human, Mouse, Rat
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P62166
  • P62166
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Neuronal calcium sensor 1 protein (1-190AA)
  • Recombinant Human Neuronal calcium sensor 1 protein (1-190AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together