14009
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-586
- 77739-216
- Antigen symbol:
- GTF2E2
- Antigen synonyms:
- H1 histone family member 2|H1.a|H12_HUMAN|H1F2|H1s-1|HIST1H1C|Histone 1 H1c|Histone cluster 1 H1c|Histone H1.2|Histone H1c|Histone H1d|Histone H1s-1|MGC3992
- FE|General transcription factor IIE subunit 2|General transcription factor IIE|polypeptide 2|beta 34kDa|gtf2e2|T2EB_HUMAN|TF2E 2|TF2E2|TFIIE B|TFIIE-beta|Transcription initiation factor IIE beta subunit|Transcription initiation factor IIE subunit beta
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- P16403
- P29084
- Isotype:
- IgG
- Western blot:
- Yes
- ELISA:
- Yes
- Immunogen:
- Peptide sequence around site of Lys (186) derived from Human Histone H1.2
- Recombinant Human Transcription initiation factor IIE subunit beta protein (1-291AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together