Anti-METAP1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: METAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-674
- 77743-022
- Antigen symbol:
- METAP1
- METAP1
- Antigen synonyms:
- AMPM1_HUMAN|DKFZp781C0419|KIAA0094|MAP 1|MAP1A|MetAP 1|MetAP1A|Methionine aminopeptidase 1|Peptidase M 1
- AMPM1_HUMAN|DKFZp781C0419|KIAA0094|MAP 1|MAP1A|MetAP 1|MetAP1A|Methionine aminopeptidase 1|Peptidase M 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53582
- P53582
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Methionine aminopeptidase 1 protein (54-125AA)
- Recombinant Human Methionine aminopeptidase 1 protein (54-125AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together