Anti-LRRFIP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: LRRFIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77742-381
- 77859-159
- Antigen symbol:
- LRRFIP1
- LRRFIP1
- Antigen synonyms:
- FLAP 1|FLAP1|FLIIAP 1|FLIIAP1|GC binding factor 2|GC-binding factor 2|GCF 2|GCF2|HUFI 1|HUFI1|Leucine rich repeat (in FLII) interacting protein 1|Leucine rich repeat flightless interacting protein 1|Leucine-rich repeat flightless-interacting protein 1|LRR FLII interacting protein 1|LRR FLII-interacting protein 1|LRRF1_HUMAN|LRRFIP 1|Lrrfip1|MGC10947|MGC119738|MGC119739|NEDD8 conjugating enzyme|TAR RNA interacting protein|TAR RNA-interacting protein|Transcription factor 9 like|TRIP
- FLAP 1|FLAP1|FLIIAP 1|FLIIAP1|GC binding factor 2|GC-binding factor 2|GCF 2|GCF2|HUFI 1|HUFI1|Leucine rich repeat (in FLII) interacting protein 1|Leucine rich repeat flightless interacting protein 1|Leucine-rich repeat flightless-interacting protein 1|LRR FLII interacting protein 1|LRR FLII-interacting protein 1|LRRF1_HUMAN|LRRFIP 1|Lrrfip1|MGC10947|MGC119738|MGC119739|NEDD8 conjugating enzyme|TAR RNA interacting protein|TAR RNA-interacting protein|Transcription factor 9 like|TRIP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q32MZ4
- Q32MZ4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leucine-rich repeat flightless-interacting protein 1 protein (1-315AA)
- Recombinant Human Leucine-rich repeat flightless-interacting protein 1 protein (1-315AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together