Anti-OR51A4 Rabbit Polyclonal Antibody
Catalog # 77738-917
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:OR51A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8NGJ6
- Antigen synonyms:Olfactory receptor 51A4
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Olfactory receptor 51A4
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA: 1:10000
Type:
Antigen: OR51A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human