Anti-FKTN Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FKTN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-259
- 77855-450
- Antigen symbol:
- FKTN
- FKTN
- Antigen synonyms:
- FCMD gene|Fukuyama type congenital muscular dystrophy protein|FKTN_HUMAN|MDDGC4|MDDGA4|MDDGB4|MGC126857|LGMD2M|OTTHUMP00000021841|FCMD|MGC138243|CMD1X|Fukutin|patient fukutin|Fukuyama-type congenital muscular dystrophy protein|MGC134944|MGC134945
- CMD1X|FCMD|FCMD gene|FKTN_HUMAN|Fukutin|Fukuyama type congenital muscular dystrophy protein|Fukuyama-type congenital muscular dystrophy protein|LGMD2M|MDDGA4|MDDGB4|MDDGC4|MGC126857|MGC134944|MGC134945|MGC138243|OTTHUMP00000021841|patient fukutin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75072
- O75072
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fukutin protein (87-276AA)
- Recombinant Human Fukutin protein (87-276AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together